Anti-IL15RA Goat Polyclonal Antibody

Supplier: R&D Systems
AF247
220010-540EA 750.55 USD
220010-540
Anti-IL15RA Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL15RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR