Anti-CD69 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2386
220010-483EA 707.17 USD
220010-483
Anti-CD69 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD69
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR