Anti-IL6R Goat Polyclonal Antibody

Supplier: R&D Systems
AF-227-NA
220010-417EA 736.09 USD
220010-417
Anti-IL6R Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL6R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR