Anti-CA2 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF2184
220010-353EA 663.78 USD
220010-353
Anti-CA2 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR