Anti-HC Goat Polyclonal Antibody

Supplier: R&D Systems
AF2150
220010-335EA 721.63 USD
220010-335
Anti-HC Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: HC
Antigen Name: hemolytic complement
Antigen Synonyms: C5, C5a, He
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR