Anti-CSF3 Goat Polyclonal Antibody

Supplier: R&D Systems
AF-214-NA
220010-334EA 736.09 USD
220010-334
Anti-CSF3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR