Anti-SIGLEC10 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2130
220010-328EA 707.17 USD
220010-328
Anti-SIGLEC10 Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: SIGLEC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR