Anti-SEMA6B Goat Polyclonal Antibody

Supplier: R&D Systems
AF2094
220010-304EA 678.25 USD
220010-304
Anti-SEMA6B Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SEMA6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR