Anti-C1S Sheep Polyclonal Antibody

Supplier: R&D Systems
AF2060
220010-281EA 678.25 USD
220010-281
Anti-C1S Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C1S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR