Anti-CD83 Goat Polyclonal Antibody

Supplier: R&D Systems
AF2044
220010-266EA 634.86 USD
220010-266
Anti-CD83 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR