Anti-PRKRA Goat Polyclonal Antibody

Supplier: R&D Systems
AF1980
220010-228EA 649.32 USD
220010-228
Anti-PRKRA Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRKRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR