Anti-DHH Goat Polyclonal Antibody

Supplier: R&D Systems
AF196
220010-216EA 678.25 USD
220010-216
Anti-DHH Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DHH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR