Anti-MAPK9 Rabbit Polyclonal Antibody

Supplier: R&D Systems
AF1846
220010-150EA 577.02 USD
220010-150
Anti-MAPK9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAPK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR