Anti-CDH11 Goat Polyclonal Antibody

Supplier: R&D Systems
AF1790
220010-119EA 692.71 USD
220010-119
Anti-CDH11 Goat Polyclonal Antibody
Antibodies
Adhesion Blockade

Type: Primary
Antigen: CDH11
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR