Anti-AKT2 Rabbit Polyclonal Antibody

Supplier: R&D Systems
AF1775
220010-109EA 577.02 USD
220010-109
Anti-AKT2 Rabbit Polyclonal Antibody
Antibodies
Simple Western

Type: Primary
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR