Anti-MAPK12 Goat Polyclonal Antibody

Supplier: R&D Systems
AF1644
220010-033EA 678.25 USD
220010-033
Anti-MAPK12 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAPK12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR