Anti-CD276 Goat Polyclonal Antibody

Supplier: R&D Systems
AF1397
220009-885EA 692.71 USD
220009-885
Anti-CD276 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD276
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR