Anti-KNG1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF1396
220009-884EA 707.17 USD
220009-884
Anti-KNG1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR