Anti-EPOR Goat Polyclonal Antibody

Supplier: R&D Systems
AF1390
220009-882EA 678.25 USD
220009-882
Anti-EPOR Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EPOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR