Anti-SELP Sheep Polyclonal Antibody

Supplier: R&D Systems
AF137
220009-869EA 707.17 USD
220009-869
Anti-SELP Sheep Polyclonal Antibody
Antibodies
Adhesion Blockade

Type: Primary
Antigen: SELP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR