Anti-WIF1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF134
220009-849EA 678.25 USD
220009-849
Anti-WIF1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR