Anti-FCGR2A Goat Polyclonal Antibody

Supplier: R&D Systems
AF1330
220009-844EA 707.17 USD
220009-844
Anti-FCGR2A Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen Symbol: FCGR2A
Antigen Name: Fc fragment of IgG, low affinity IIa, receptor (CD32)
Antigen Synonyms: CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR