Anti-NTN4 Goat Polyclonal Antibody

Supplier: R&D Systems
AF1254
220009-805EA 678.25 USD
220009-805
Anti-NTN4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NTN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR