Anti-ALCAM Goat Polyclonal Antibody

Supplier: R&D Systems
AF1172
220009-749EA 721.63 USD
220009-749
Anti-ALCAM Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR