Anti-PDCD1LG2 Goat Polyclonal Antibody

Supplier: R&D Systems
AF1022
220009-638EA 678.25 USD
220009-638
Anti-PDCD1LG2 Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: PDCD1LG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR