Anti-CDH5 Goat Polyclonal Antibody

Supplier: R&D Systems
AF1002
220009-621EA 793.94 USD
220009-621
Anti-CDH5 Goat Polyclonal Antibody
Antibodies
Simple Western

Type: Primary
Antigen: CDH5
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR