Anti-CSF2 Goat Polyclonal Antibody

Supplier: R&D Systems
AB-415-NA
220009-596EA 577.02 USD
220009-596
Anti-CSF2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR