Anti-IL1RN Goat Polyclonal Antibody

Supplier: R&D Systems
AB-280-NA
220009-569EA 692.71 USD
220009-569
Anti-IL1RN Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL1RN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR