Anti-TGF-beta 5 Goat Polyclonal Antibody

Supplier: R&D Systems
AB-245-NA
220009-548EA 765.02 USD
220009-548
Anti-TGF-beta 5 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: TGF-beta 5
Antigen Name:
Antigen Synonyms:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR