Anti-GDNF Goat Polyclonal Antibody

Supplier: R&D Systems
AB-212-NA
220009-525EA 736.09 USD
220009-525
Anti-GDNF Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GDNF
Antigen Name: glial cell derived neurotrophic factor
Antigen Synonyms: ATF1, ATF2, HFB1-GDNF, HSCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR