Anti-CXCL8 Goat Polyclonal Antibody

Supplier: R&D Systems
AB-208-NA
220009-520EA 736.09 USD
220009-520
Anti-CXCL8 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR