SMD-VAC™-HP Vacuum Handling System, Virtual Industries
Supplier: Virtual Industries
VFS-1
21925-200EA 347.53 USD
21925-200
SMD-VAC™-HP Vacuum Handling System, Virtual Industries
Vacuum Handling Systems
Made with extra power to pick up very small to very large-size components, the SMD-VAC™-HP features a precision pump and motor that generates over 15"Hg
Compact tower design has a small footprint. Electric eye pen holder automatically shuts the unit down when pen is returned to holder. Optional foot switch 21925-200 controls vacuum during pick-and-place operations.
Ordering information: Complete with 1.2m (4') of coiled and straight vacuum hose, a vacuum pick-up handle, set of seven vacuum tips, 12V DC wall transformer, automatic pump on/off control, rubber nonskid feet and ESD connection.