COTTON FILTER F/GS80/80I
Supplier: NILFISK-ADVANCED AMERICA
Napped Cotton Filter^ For the second stage of filtration in the Nilfisk vacuum models GM-80, GM-80CR, and GM-811. Retains 99.8% of particles to 3 microns in size. Over-sized to provide maximum filter surface area.