Wet/Dry HEPA Vacuum, Nilfisk-Advance America
Supplier: Nilfisk
01699564
21925-028EA 12209.8 USD
21925-028
Wet/Dry HEPA Vacuum, Nilfisk-Advance America
Vacuum Cleaners
Ideal for cleanroom environments, the VT 60CR vacuum features a specially designed system that prevents motor carbon dust from escaping through the exhaust system
It cleans up wet or dry surfaces, with no filter change needed. Main filter consists of 105 porous polyethylene tubes that provide excellent resistance to water, mildew, rot, and corrosion. Internal cotton microfilter and HEPA filter retain 99.97% of all ultrafine particles down to and including 0.3µm. Initial foam impact filter for coarse, wet materials protects float valve from clogging. Pliable inlet tube aids in releasing damp, caking debris. Total filter area (including impact filter, filter finger tubes, microfilter, and HEPA or ULPA cartridge) is 14,290cm2 (2215in.2). Motor is grounded.
Ordering information: Supplied with trolley, 3m (10') plastic hose, double-curved aluminum wand, 35.6cm (14") wet floor nozzle, 7.6cm (3") aluminum dust brush, 27.9cm (11") plastic crevice tool, 10m (30') power cord and twenty-five 37.9L (10gal.) polyliners. Replacement cotton microfilter retains 99.5% of particulates at 2µm. Optional ULPA filter 21925-021 retains 99.999% of 0.12µm particles. Tool box accessory 21925-022 mounts on trolley to store vacuum parts.