VWR® Spec-Wipe® 3 Sterile Wiper
Supplier: VWR International
21914-777
21914-777CS 621.62 USD
21914-777
VWR® Spec-Wipe® 3 Sterile Wiper
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Sterile wiper has superior quality for aseptic environments where bacteria, particulate, and extractable levels are critical concerns.
  • Hydroentangled
  • Mechanically bonded
  • Minimize fiber and particle generation
Made of highly absorbent 45% polyester and 55% cellulose blend.
Wipers are mechanically bonded without the use of chemicals and sterilized by gamma irradiation to a Sterility Assurance Level of 10⁻⁶ in accordance with ANSI/ISO/AAMI guidelines.
Packaging: Packaged in 25 inner bags of 20 wipers.