SatPax® 3000, Presaturated Wipers, Berkshire
Supplier: Berkshire
SPX300000112
21912-110CS 1067.12 USD
21912-110
SatPax® 3000, Presaturated Wipers, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Berkshire SatPax® 3000 combines the UltraSeal® 3000 sealed-edge, cleanroom-laundered wiper (with two-ply pinsonic tubular construction) with various concentrations of isopropyl alcohol (IPA)- the SPX3000.001.12 contains a 70% IPA solution and the SPX3000.002.12 contains a 9% IPA solution.
  • Critically low particles, fibers, ions, and extractables
  • Patented sealed edge process combined with tubular design offers excellent abrasion resistance for the toughest cleaning applications
  • Pinsonic design adds surface texture that aids in stubborn residue removal
  • Reduces alcohol usage and preparation/handling costs
  • Reduces VOC emissions
  • Increases cleaning efficiency and cleaning protocol consistency
This pre-wetted format provides a more cost-effective and easy-to-use solution compared to the manual wetting of dry wipers, which requires bulk handling of messy solvents and maintenance of squirt bottles and results in inconsistent wetting and cleaning.
The cleanroom wiper is composed of 100% continuous filament polyester knit fabric.