VWR® Spec-Wipe® 4 Wiper
Supplier: VWR International
21912-046
21912-046CS 831.12 USD
21912-046
VWR® Spec-Wipe® 4 Wiper
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Made of 100% double-knit polyester, delivering softness, high absorbency, strength, and abrasion resistance.
  • Textured yarn construction
  • Clean-cut edges
  • Proprietary cleaning process reduces particulates and extractable contaminants
Continuous-filament construction fortifies the fabric against breakdown.
Textured yarn construction and clean-cut edges result in less linting and snagging. Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Certifications: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Packaging: Wipes are flat-packed, 8 packs of 150/case (1200 wipers per case).