Sporicidin® Enzymatic Cleaner, Contec®
Supplier: Contec
ENZ-1284
21909-840CS 340.45 USD
21909-840
Sporicidin® Enzymatic Cleaner, Contec®
Cleaning Agents Detergents and Surface Cleansers
Sporicidin® enzymatic cleaner is a blend of enzymes in a low foaming solution formulated to loosen and remove dried debris, protein, blood, and mucin deposits. It effectively cleans both natural and synthetic fibers that have been soiled with organic and inorganic debris.
  • Special blend contains cellulose and protein enzymes for cleaning and removing material stains
  • Breaks down both organic and inorganic debris
  • Mildly alkaline (pH 7.5) and contains enzyme preservative for added stability
  • Low foaming, concentrated formulation
  • Safe for use on rubber, stainless steel, vinyl, plastics, linens, clothing, carpets, fabrics and upholstery
  • Reduces the need for manual scrubbing
  • Non-flammable, non-staining, and non-toxic
Sporicidin® Enzymatic Cleaner can be used in ultrasonic units, automatic washers, and evacuation systems. This highly effective cleaner and stain remover has a neutral pH and is biodegradable, nonflammable and nontoxic. It is ideal for household, medical, dental, institutional, healthcare, veterinary, and cleanroom use.
Economical, highly concentrated formula:
1 ounce makes 1 gallon of cleaning solution
1 gallon (3.8 l) of concentrate makes 128 gallons (484.5 l) of solution
55 gallons (208.2 l) of concentrate makes 7040 gallons (26649 l) of solution