Olsen-Hegar Needle Holders, Integra™ Miltex®
Supplier: Miltex
8-14 8-15 8-16TC 8-15TC 8-16
21909-356EA 343.01 USD
21909-356 21909-358 21909-176 11000-482 11000-484
Olsen-Hegar Needle Holders, Integra™ Miltex®
Forceps
Needle Holders with Suture Scissors
A close examination of Miltex Needle Holders, either Standard or Tungsten Carbide, will reveal meticulously crafted surgical tools that will perform precisely as intended. Smoothly beveled box locks and jaw edges protect delicate suture material from snags or cutting. The jaw surface is milled to ensure a secure grasp and to prevent needle slippage or rotation.