VWR® Cotton Pads
Supplier: VWR International
21902-985
21902-985CS 1114 USD
21902-985
VWR® Cotton Pads
Wipes Reusable Cleaning Cloths Cloths for General Applications
Soft, pure, highly absorbent pads will not scratch the most delicate coatings or surfaces.
  • No Binders or chemicals
  • Excellent wet strength
Economical, all-purpose 100% pure cotton pads. Useful for the printing industry. Will not contaminate developing solutions or chemicals.