VWR® Digital Counter with Keychain, Wrist Strap, and Carabiner
Supplier: VWR International
90027-45
21800-062EA 34.9 USD
21800-062
VWR® Digital Counter with Keychain, Wrist Strap, and Carabiner
Cash and Check Handling Supplies Tally Counters
Designed for ease-of-use and fail-safe counting, this lightweight digital counter features a water-resistant O-ring seal, five-digit display count, button click for count confirmation, and recessed reset to prevent accidental tampering.
  • Easy-to-use design for fail-safe counting
  • Water-resistant O-ring seal
  • Recessed reset to prevent accidental tampering
Ordering information: Supplied with two-year constant-use battery and shock-resistant ABS plastic case.