VWR® Conductivity/Temperature Meter
Supplier: VWR International
21800-012
21800-012EA 1187.89 USD
21800-012
VWR® Conductivity/Temperature Meter
Electrochemistry Meters Conductivity Meters
Two displays simultaneously show conductivity reading and temperature measurements on 34.9 mm (1³/₈") LCD readout.
  • Serial RS232 output allows meter to be connected to a computer or data logger for monitoring and keeping results
  • Recall highest, lowest, and average readings at the touch of a button
  • 'Data Hold' button freezes the display to capture readings
Meter has a controller to adjust a probe’s K (constant) factor and to calibrate to solution standards.
Data acquisition system (61161-386) captures, displays, and stores readings on any PC. Information can be imported into databases.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Meter is supplied with probe with 101.6 cm (40") cable, serial computer output, and 9V alkaline battery. Data acquisition system includes a 3¹/₂" diskette and a 91.4 cm (36") serial cable with D9F plug.