VWR® Talon® Double Jaw Utility Clamp
Supplier: VWR International
21573-800
21573-800EA 68.5 USD
21573-800
VWR® Talon® Double Jaw Utility Clamp
Clamps General Purpose Clamps
The Talon® Double Jaw Utility Clamp holds two sizes of object with care and features a medium and a large two-prong clamp that swivel 360° in one plane.
  • Double jaw design holds two objects simultaneously
  • Vinyl sleeves
  • Fiberglass covers for high-temperature applications
The medium clamp has a maximum opening of 76 mm (3"). The large clamp has a maximum opening of 95 mm (33/4"). Supplied with vinyl sleeves, and fiberglass covers for temperatures above 100°C (212°F).