VWR® Talon® Wall Clamp
Supplier: VWR International
21573-154
21573-154CS 2548.14 USD
21573-154
VWR® Talon® Wall Clamp
Clamps General Purpose Clamps
Clamp is designed for securing fermentation tubes, burets, or other small objects where frames are not available.
  • Secure holding of small objects
  • Integral self-tapping screw
  • Nickel-plated construction
Integral self-tapping screw secures clamp to wall.