VWR® Talon® Suspension Clamp
Supplier: VWR International
21573-153
21573-153CS 1139.66 USD
21573-153
VWR® Talon® Suspension Clamp
Clamps General Purpose Clamps
Clamp holds thermometers, potash bulbs, or drying tubes 114 mm (41/2") from the support rod.
  • Machine-tapered hook
  • Nickel-plated brass construction
  • Built-in holder
Machine-tapered, nickel-plated brass hook won't bend, rust, or corrode. Built-in holder grips rods up to 19 mm (3/4") in diameter.