VWR® Talon® All-Position Clamp Holder
Supplier: VWR International
21572-658
21572-658CS 464.7 USD
21572-658
VWR® Talon® All-Position Clamp Holder
Clamps Bossheads
Surpass standard holding capabilities with this all-position clamp holder.
  • Allows 360° rotation
  • Fully adjustable holder
Building on the upgraded Q-clamp design, the holder allows adjustment at any angle in any plane. Holders are set at 90° to each other connected by a 90° connector for 360° rotation.