WHEATON® Diazomethane Generator with ST Clear-Seal™ Grease-Free Joints, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
281135
21518-132CS 160.84 USD
21518-132
WHEATON® Diazomethane Generator with ST Clear-Seal™ Grease-Free Joints, DWK Life Sciences
Gas Generators Compressed Air Stations
This Diazomethane Generator is used to generate diazomethane and other gases for gas chromatography, mass spectroscopy and NMR studies using one millimole of Methylnitronitrosoguanidine (MNNG).
  • Clear-Seal™ grease-free joints that have never been ground and does not require an 'O' ring
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
Includes a plastic standard taper joint clamp, a 13-425 screw cap with cut-out top and a PTFE faced rubber liner.
Caution: Please use a safety shield and efficient fume hood when generating diazomethane or similar gases.