Shut-Off Valves, Upchurch Scientific®
Supplier: Upchurch Scientific
P-783
21511-232EA 80.17 USD
21511-232
Shut-Off Valves, Upchurch Scientific®
Valves Non Return Valves
Stop a flow stream quickly with these shut-off valves. The bodies are manufactured from either PEEK or ETFE and both versions feature a PCTFE rotor, making them highly resistant to chemicals. The blue colorant used in some valve configurations has proven not to leach out with common HPLC solvents.
  • For low and medium pressure applications
  • Biocompatible, all-polymer flow path
  • Pressure rated to 500 psi (34 bar)