Tubing Cutters, Plastic
Supplier: Optimize
10-10-00139
21509-054EA 92.8 USD
21509-054
Tubing Cutters, Plastic
Cutters
These plastic tubing cutters makes clean, burr-free, right-angled cuts on polymeric tubing. Versatile and easy to use, these can be used with a variety of ODs.
  • Produces defined, sharp edges when used on polymeric tubing
  • Versatile and user-friendly
  • For use with various ODs