PYREX® Chromatography Column with Coarse Fritted Disc, Corning
Supplier: Corning
38450-10 38450-20 38450-40
89091-058CS 858.41 USD
89091-058 89091-236 21506-063
PYREX® Chromatography Column with Coarse Fritted Disc, Corning
Chromatography Columns
These columns are designed for general chromatography analysis.
The tops are tooled for rubber stopper fit and the tubes are fabricated with a sealed-in coarse porosity fritted disc.